Oxythiamine · HCl

Supplier: BACHEM AMERICAS INC

4002126.0025 4002126.0100
Q-1365.0025BAEA 1966.97 CAD
Q-1365.0025BA Q-1365.0100BA
Oxythiamine · HCl
Oxythiamine · HCl

Sequence: Oxythiamine · HCl


Thiamine antagonist and inhibitor of saccharomyces cerevisiae transketolase. Raïs et al. indicated that oxythiamine inhibits the growth of Ehrlich's tumor cells.

Formula: C₁₂H₁₆ClN₃O₂S·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00050656
CAS Number: 614-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR