4-Methoxy-2-naphthylamine

Supplier: BACHEM AMERICAS INC

Synonyms: 4-Methoxy-β-naphthylamine

4001634.0250 4001634.1000
Q-1315.0250BAEA 258.14 CAD
Q-1315.0250BA Q-1315.1000BA
4-Methoxy-2-naphthylamine
4-Methoxy-2-naphthylamine

Sequence: 4-Methoxy-β-naphthylamine
Synonym(s): 4-Methoxy-2-naphthylamine


Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").


Fluorescence standard for assays, cleavage product of 4MβNA (MNA) substrates.

Formula: C₁₁H₁₁NO
MW: 173.215 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00056592
CAS Number: 2764-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR