Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

4006805.0025 4006805.0100 4006805.0250
Q-1240.0025BAEA 428.53 CAD
Q-1240.0025BA Q-1240.0100BA Q-1240.0250BA
Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimide)

Sequence: Fmoc-OSu
Synonym(s): N-(9-Fluorenylmethyloxycarbonyloxy)-succinimide


Most efficient reagent for the selective preparation of Fmoc-derivatives of hydroxy amino acids in high yield. The advantage of Fmoc-OSu is that little or no oligopeptides are formed when amino acid derivatives are prepared. The use of this reagent in the synthesis of glycopeptides has been reported as well.

Formula: C₁₉H₁₅NO₅
MW: 337.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010733
CAS Number: 82911-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR