Dithiothreitol (DTT, Cleland's reagent)

Supplier: BACHEM AMERICAS INC

Synonyms: DL-1,4-DithiothreitolDTTcis-2,3-Dihydroxy-1,4-butanedithiol(R*,R*)-1,4-Dimercapto-2,3-butandiol

4031235.0025
Q-1225.0025BAEA 521.48 CAD
Q-1225.0025BA
Dithiothreitol (DTT, Cleland's reagent)
Dithiothreitol (DTT, Cleland's reagent)

Sequence: Threo-1,4-dimercapto-2,3-butanediol
Synonym(s): Cleland's Reagent#DTT#Dithiothreitol


For DOTA(OtBu)₃ see Q-2525.


Reagent for quantitative reduction of disulfide groups. Hygroscopic. DTT has been employed as substrate in combination with homovanillic acid in an continuous fluorescence assay for sulfhydryl oxidase.

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://ca.vwr.com/stibo/web/std.lang.all/98/19/2299819.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR