N-(Benzyloxycarbonyloxy)succinimide

Supplier: BACHEM AMERICAS INC

4031237.0100 4031237.0500
Q-1160.0100BAEA 238.11 CAD
Q-1160.0100BA Q-1160.0500BA
N-(Benzyloxycarbonyloxy)succinimide
N-(Benzyloxycarbonyloxy)succinimide

Sequence: Z-OSu
Synonym(s): N-(Benzyloxycarbonyloxy)succinimide


Reagent for introducing the Z-amino protecting group.

Formula: C₁₂H₁₁NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00005513
CAS Number: 13139-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR