Anisindione

Supplier: BACHEM AMERICAS INC

4028913.0005 4028913.0025
Q-1085.0005BAEA 1041.05 CAD
Q-1085.0005BA Q-1085.0025BA
Anisindione
Anisindione

Sequence: Anisindione
Synonym(s): 2-p-Anisyl-1,3-indandione


Anisindione is a synthetic anticoagulant preventing the formation of active procoagulation factors II, VII, IX, and X, as well as the anticoagulant proteins C and S, in the liver by inhibiting the vitamin K-mediated gamma-carboxylation of precursor proteins.

Formula: C₁₆H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00176194
CAS Number: 117-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR