4-Aminophenyl-1-thio-β-D-galactopyranoside
Supplier: BACHEM AMERICAS INC
Synonyms:
4-Aminophenyl-1-thio-β-D-galactopyranoside
4014175.0250
4014175.1000
Q-1070.0250BAEA
516.34
CAD
Q-1070.0250BA
Q-1070.1000BA
4-Aminophenyl-1-thio-β-D-galactopyranoside
4-Aminophenyl-1-thio-beta-D-galactopyranoside
Sequence: 4-Aminophenyl-1-thio-β-D-galactopyranoside
See also the indigogenic β-galactosidase substrates M-1190 (X-Gal) and Q-1270.
Inhibitor of E.coli β-galactosidase
Formula:
C₁₂H₁₇NO₅S MW: 287.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00036767 CAS Number: 29558-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...