N-Acetylmuramic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 2-Acetamido-2-deoxy-3-O-(d-1-carboxyethyl)-D-glucopyranose, 2-Acetamido-2-deoxy-3-O-(D-1-carboxyethyl)-D-glucopyranose, (R)-2-(((2R,3R,4R,5R)-2-Acetamido-4,5,6-trihydroxy-1-oxohexan-3-yl)oxy)propanoic acid, MurNAc, NAMA

4011440.0100 4011440.1000
Q-1015.0100BAEA 211.77 CAD
Q-1015.0100BA Q-1015.1000BA
N-Acetylmuramic acid
N-Acetylmuramic acid

Sequence: Ac-muramic acid
Synonym(s): MurNAc#NAMA


A component of bacterial cell walls. For a protected derivative of MurNAc see Q-1005.

Formula: C₁₁H₁₉NO₈
MW: 293.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00221511
CAS Number: 10597-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR