N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)

Supplier: BACHEM AMERICAS INC

4058964.0001 4058964.0025
P-1165.0001BAEA 180.91 CAD
P-1165.0001BA P-1165.0025BA
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)
N-Methyl-N-nonanoyl-D-glucamine (MEGA-9)

Sequence: MEGA-9
Synonym(s): Nonanoyl-N-methylglucamide


Non-ionic detergents with high solubilizing power and nondenaturing properties. They can easily be removed by dialysis. Bachem offers MEGA-8, -9, -10, and -12.

Formula: C₁₆H₃₃NO₆
Storage Temperature: Freezer
CAS Number: 85261-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR