Pentaethylene glycol monododecyl ether
Supplier: BACHEM AMERICAS INC
Synonyms:
Dodecyl pentaethylene glycolether
4028288.0005
4028288.0025
4028288.0001
P-1160.0005BAEA
828.77
CAD
P-1160.0005BA
P-1160.0025BA
P-1160.0001BA
Pentaethylene glycol monododecyl ether
Pentaethylene glycol monododecyl ether
Dodecyl pentaethylene glycolether
CAS: 3055-95-6
Formula: C22H46O6
M.W: 406.6
Storage temp.: -20 ± 5 °C
Formula:
C₂₂H₄₆O₆ MW: 406.60 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00043064 CAS Number: 3055-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...