N-Octylpolyoxyethylene

Supplier: BACHEM AMERICAS INC

Synonyms: Rosenbusch-Tenside, Octyl-POE

4031122.0025 4031122.0100 4031122.0250
P-1140.0025BAEA 683.51 CAD
P-1140.0025BA P-1140.0100BA P-1140.0250BA
N-Octylpolyoxyethylene
N-Octylpolyoxyethylene

Octyl-POE solubilizes membrane proteins. The CMC value is 6.6 mM.


Octyl-POE solubilizes membrane proteins. The CMC value is 6.6 mM.

Storage Temperature: Refrigerator
MDL Number: MFCD00801422
CAS Number: 27252-75-1
EINECS: 500-058-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR