Tetraethylene glycol monooctyl ether

Supplier: BACHEM AMERICAS INC

4006356.0005 4006356.0025 4006356.0100
P-1120.0005BAEA 407.1 CAD
P-1120.0005BA P-1120.0025BA P-1120.0100BA
Tetraethylene glycol monooctyl ether
Tetraethylene glycol monooctyl ether

Sequence: n-Octyltetraoxyethylene
Synonym(s): C₈E₄#Tetraethyleneglycolmonooctylether


The tenside C8E4 solubilizes membrane proteins. The CMC value is 7.2 mM.

Formula: C₁₆H₃₄O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00043033
CAS Number: 19327-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR