N-Octyl-β-D-glucopyranoside

Supplier: BACHEM AMERICAS INC

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

4005687.0005 4005687.0025 4005687.0001
P-1110.0005BAEA 871.29 CAD
P-1110.0005BA P-1110.0025BA P-1110.0001BA
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside

Octyl glucoside
CAS: 29836-26-8
Formula: C14H28O6
M.W: 292.37
Storage temp.: -20 ± 5 °C

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR