Triethylene glycol monohexyl ether

Supplier: BACHEM AMERICAS INC

Synonyms: n-Hexyltrioxyethylene

4004291.0005 4004291.0025
P-1095.0005BAEA 387.21 CAD
P-1095.0005BA P-1095.0025BA
Triethylene glycol monohexyl ether
Triethylene glycol monohexyl ether

Sequence: n-Hexyltrioxyethylene
Synonym(s): C₆E₃#Triethyleneglycolmonohexylether

Formula: C₁₂H₂₆O₄
MW: 234.336 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00056659
CAS Number: 25961-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR