3,6,9,12,15-Pentaoxapentacosan-1-ol

Supplier: BACHEM AMERICAS INC

Synonyms: n-Decylpentaoxyethylene

4006590.0005 4006590.0025
P-1005.0005BAEA 774.28 CAD
P-1005.0005BA P-1005.0025BA
3,6,9,12,15-Pentaoxapentacosan-1-ol
3,6,9,12,15-Pentaoxapentacosan-1-ol

Sequence: n-Decylpentaoxyethylene
Synonym(s): C₁₀E₅#Pentaethyleneglycolmonodecylether

Formula: C₂₀H₄₂O₆
MW: 378.55 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00043363
CAS Number: 23244-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR