N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

Supplier: BACHEM AMERICAS INC

4011133.0005 4011133.0025 4011133.0001
P-1000.0005BAEA 746.95 CAD
P-1000.0005BA P-1000.0025BA P-1000.0001BA
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

MEGA-10
CAS: 85261-20-7
Formula: C17H35NO6
M.W: 349.47
Storage temp.: -20 ± 5 °C

Formula: C₁₇H₃₅NO₆
Storage Temperature: Freezer
MDL Number: MFCD00036801
CAS Number: 85261-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR