1,3-Dipalmitoylglycero-2-phosphoethanolamine

Supplier: BACHEM AMERICAS INC

4040080.0100 4040080.0500
O-1605.0100BAEA 387.21 CAD
O-1605.0100BA O-1605.0500BA
1,3-Dipalmitoylglycero-2-phosphoethanolamine
1,3-Dipalmitoylglycero-2-phosphoethanolamine

Sequence: 1,3-Dipalmitoyl-glycero-2-phosphoethanolamine

Formula: C₃₇H₇₄NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00038196
CAS Number: 67303-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR