SN-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2,3-Dihydroxypropyl (2-(trimethylammonio)ethyl) phosphate

4030680.0025 4030680.0100
O-1590.0025BAEA 434.83 CAD
O-1590.0025BA O-1590.0100BA
SN-glycero-3-phosphocholine
SN-glycero-3-phosphocholine

Sequence: sn-Glycero-3-phosphocholine
Synonym(s): Choline alfoscerate#L-α-GPC#L-α-Lecithin


Intermediate for the synthesis of phosphatidylcholines, acetylcholine precursor, and muscarinic receptor agonist. Choline alphoscerate has shown to be effective in countering cognitive symptoms in forms of dementia disorders of degenerative, vascular or combined origin.

Formula: C₈H₂₀NO₆P
Storage Temperature: Freezer
MDL Number: MFCD00063544
CAS Number: 28319-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR