1,2-Didodecanoyl-rac-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

4018411.0250 4018411.1000
O-1510.0250BAEA 206.51 CAD
O-1510.0250BA O-1510.1000BA
1,2-Didodecanoyl-rac-glycero-3-phosphocholine
1,2-Didodecanoyl-rac-glycero-3-phosphocholine

Sequence: 1,2-Dilauroyl-rac-glycero-3-phosphocholine
Synonym(s): DL-β,γ-Dilauroyl-α-lecithin

Formula: C₃₂H₆₄NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00043210
CAS Number: 18656-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR