1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

Synonyms: L-β-Oleoyl-γ-palmitoyl-α-lecithin

4004281.0100 4004281.1000
O-1410.0100BAEA 387.21 CAD
O-1410.0100BA O-1410.1000BA
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine

Sequence: L-β-Oleoyl-γ-palmitoyl-α-lecithin
Synonym(s): POPC#1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine

Formula: C₄₂H₈₂NO₈P
MW: 760.091 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077387
CAS Number: 26853-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR