Batyl alcohol

Supplier: BACHEM AMERICAS INC

4014011.0005 4014011.0025
O-1370.0005BAEA 665.15 CAD
O-1370.0005BA O-1370.0025BA
Batyl alcohol
Batyl alcohol

Sequence: 1-O-Octadecyl-rac-glycerol
Synonym(s): Batyl alcohol#Batilol

Formula: C₂₁H₄₄O₃
Storage Temperature: Freezer
MDL Number: MFCD00004720
CAS Number: 544-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR