1-O-Octadecyl-sn-glycerol

Supplier: BACHEM AMERICAS INC

4010401.0001 4010401.0005
O-1365.0001BAEA 413.58 CAD
O-1365.0001BA O-1365.0005BA
1-O-Octadecyl-sn-glycerol
1-O-Octadecyl-sn-glycerol

Sequence: 1-O-Octadecyl-sn-glycerol

Formula: C₂₁H₄₄O₃
Storage Temperature: Freezer
MDL Number: MFCD00237201
CAS Number: 6129-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR