Chimyl alcohol

Supplier: BACHEM AMERICAS INC

4010402.0001 4010402.0005
O-1285.0001BAEA 423.95 CAD
O-1285.0001BA O-1285.0005BA
Chimyl alcohol
Chimyl alcohol

Sequence: 1-O-Hexadecyl-sn-glycerol

Formula: C₁₉H₄₀O₃
Storage Temperature: Freezer
MDL Number: MFCD00151171
CAS Number: 506-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR