1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

4006552.0025 4006552.0100 4006552.0500
O-1270.0025BAEA 402.08 CAD
O-1270.0025BA O-1270.0100BA O-1270.0500BA
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine

Sequence: 1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine


O-1270 corresponds to PAF (C₁₆), O-1355 to PAF (C₁₈).


Synonym(s): PAF (C₁₆)

Formula: C₂₆H₅₄NO₇P
Storage Temperature: Freezer
MDL Number: MFCD00043214
CAS Number: 74389-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR