1,2-Distearoyllecithin
Supplier: BACHEM AMERICAS INC
Synonyms:
1,2-Distearoyl-rac-glycero-3-phosphocholine
4005624.0250
4005624.1000
O-1240.0250BAEA
858.7
CAD
O-1240.0250BA
O-1240.1000BA
1,2-Distearoyllecithin
1,2-Distearoyllecithin
Sequence: 1,2-Distearoyl-rac-glycero-3-phosphocholine
Synonym(s): DL-β,γ-Distearoyl-α-lecithin
Formula:
C₄₄H₈₈NO₈P MW: 790.161 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00067532 CAS Number: 816-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...