1,2-Distearoyl-sn-glycero-3-phosphocholine
Supplier: BACHEM AMERICAS INC
Synonyms:
DSPC, 1,2-Distearoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate
4005619.0250
4005619.1000
O-1235.0250BAEA
185.3
CAD
O-1235.0250BA
O-1235.1000BA
1,2-Distearoyl-sn-glycero-3-phosphocholine
1,2-Distearoyl-sn-glycero-3-phosphocholine
Sequence: 1,2-Distearoyl-sn-glycero-3-phosphocholine
Synonym(s): DSPC#L-β,γ-Distearoyl-α-lecithin
Formula:
C₄₄H₈₈NO₈P Storage Temperature: Freezer |
MDL Number:
MFCD00036905 CAS Number: 816-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...