Distearin
Supplier: BACHEM AMERICAS INC
Synonyms:
1,2-Distearoyl-rac-glycerol
4005604.0001
4005604.0005
O-1220.0001BAEA
315.02
CAD
O-1220.0001BA
O-1220.0005BA
Distearin
Distearin
Sequence: 1,2-Distearoyl-rac-glycerol
Synonym(s): DL-α,β-Distearin
Formula:
C₃₉H₇₆O₅ MW: 625.032 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00066515 CAS Number: 1188-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...