1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

Supplier: BACHEM AMERICAS INC

4005625.0250 4005625.1000
O-1205.0250BAEA 192.25 CAD
O-1205.0250BA O-1205.1000BA
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

Sequence: 1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
Synonym(s): DPPE#L-β,γ-Dipalmitoyl-α-cephalin

Formula: C₃₇H₇₄NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00036776
CAS Number: 923-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR