1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

4004249.0250 4004249.1000
O-1195.0250BAEA 193.17 CAD
O-1195.0250BA O-1195.1000BA
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine

Sequence: 1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
Synonym(s): DPPC#L-β,γ-Dipalmitoyl-α-lecithin

Formula: C₄₀H₈₀NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00036903
CAS Number: 63-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR