1,2-Dipalmitoyl-rac-glycerol

Supplier: BACHEM AMERICAS INC

4005607.0001 4005607.0005
O-1180.0001BAEA 892.08 CAD
O-1180.0001BA O-1180.0005BA
1,2-Dipalmitoyl-rac-glycerol
1,2-Dipalmitoyl-rac-glycerol

Sequence: 1,2-Dipalmitoyl-rac-glycerol
Synonym(s): DL-α,β-Dipalmitin

Formula: C₃₅H₆₈O₅
Storage Temperature: Freezer
MDL Number: MFCD00042874
CAS Number: 40290-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR