1,2-Dipalmitoyl-sn-glycerol

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-3-Hydroxypropane-1,2-diyl dipalmitate

4005618.0001 4005618.0005
O-1175.0001BAEA 387.21 CAD
O-1175.0001BA O-1175.0005BA
1,2-Dipalmitoyl-sn-glycerol
1,2-Dipalmitoyl-sn-glycerol

Sequence: 1,2-Dipalmitoyl-sn-glycerol
Synonym(s): D-α,β-Dipalmitin

Formula: C₃₅H₆₈O₅
MW: 568.924 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00067471
CAS Number: 30334-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR