1,2-O-Dioctadecyl-sn-glycerol

Supplier: BACHEM AMERICAS INC

4005640.0001 4005640.0005
O-1155.0001BAEA 516.34 CAD
O-1155.0001BA O-1155.0005BA
1,2-O-Dioctadecyl-sn-glycerol
1,2-O-Dioctadecyl-sn-glycerol

Sequence: 1,2-O-Dioctadecyl-sn-glycerol
Synonym(s): D-α,β-Dioctadecyl-glycerol

Formula: C₃₉H₈₀O₃
Storage Temperature: Freezer
MDL Number: MFCD00236963
CAS Number: 82188-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR