Dimyristoyllecithin

Supplier: BACHEM AMERICAS INC

Synonyms: 1,2-Dimyristoyl-rac-glycero-3-phosphocholine

4006546.0250 4006546.1000
O-1140.0250BAEA 206.51 CAD
O-1140.0250BA O-1140.1000BA
Dimyristoyllecithin
Dimyristoyllecithin

Sequence: 1,2-Dimyristoyl-rac-glycero-3-phosphocholine
Synonym(s): DL-β,γ-Dimyristoyl-α-lecithin

Formula: C₃₆H₇₂NO₈P
MW: 677.945 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066562
CAS Number: 18656-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR