1,2-Dimyristoyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

Synonyms: 14:0 PC, 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, DMPC, β-γ-Dimyristoyl-L-α-Lecithin, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate

4005627.0001 4005627.0005
O-1135.0001BAEA 476.37 CAD
O-1135.0001BA O-1135.0005BA
1,2-Dimyristoyl-sn-glycero-3-phosphocholine
1,2-Dimyristoyl-sn-glycero-3-phosphocholine

Sequence: 1,2-Dimyristoyl-sn-glycero-3-phosphocholine
Synonym(s): DMPC#L-β,γ-Dimyristoyl-α-lecithin

Formula: C₃₆H₇₂NO₈P
Storage Temperature: Freezer
MDL Number: MFCD00036901
CAS Number: 18194-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR