1,2-dimyristoyl-sn-glycerol

Supplier: BACHEM AMERICAS INC

4005615.0001 4005615.0005
O-1115.0001BAEA 387.12 CAD
O-1115.0001BA O-1115.0005BA
1,2-dimyristoyl-sn-glycerol
1,2-dimyristoyl-sn-glycerol

Sequence: 1,2-Dimyristoyl-sn-glycerol
Synonym(s): D-α,β-Dimyristin

Formula: C₃₁H₆₀O₅
Storage Temperature: Freezer
MDL Number: MFCD00048454
CAS Number: 60562-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR