1,2-didodecanoyl-sn-glycerol
Supplier: BACHEM AMERICAS INC
Synonyms:
1,2-Dilauroyl-sn-glycerol
4005366.0001
4005366.0005
O-1065.0001BAEA
387.12
CAD
O-1065.0001BA
O-1065.0005BA
1,2-didodecanoyl-sn-glycerol
1,2-didodecanoyl-sn-glycerol
Sequence: 1,2-Dilauroyl-sn-glycerol
Synonym(s): D-α,β-Dilaurin
Formula:
C₂₇H₅₂O₅ MW: 456.708 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00067469 CAS Number: 60562-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...