1,2-O-Dihexadecyl-sn-glycerol

Supplier: BACHEM AMERICAS INC

4005611.0001 4005611.0005
O-1040.0001BAEA 309.81 CAD
O-1040.0001BA O-1040.0005BA
1,2-O-Dihexadecyl-sn-glycerol
1,2-O-Dihexadecyl-sn-glycerol

Sequence: 1,2-O-Dihexadecyl-sn-glycerol
Synonym(s): D-α,β-Dihexadecyl-glycerol

Formula: C₃₅H₇₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00067472
CAS Number: 67337-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR