1-O-(cis-9-Octadecenyl)-2-O-acetyl-sn-glycero-3-phosphocholine

Supplier: BACHEM AMERICAS INC

4011530.0050 4011530.0250
O-1030.0050BAEA 774.5 CAD
O-1030.0050BA O-1030.0250BA
1-O-(cis-9-Octadecenyl)-2-O-acetyl-sn-glycero-3-phosphocholine
1-O-(cis-9-Octadecenyl)-2-O-acetyl-sn-glycero-3-phosphocholine

Sequence: 1-O-(cis-9-Octadecenyl)-2-O-acetyl-sn-glycero-3-phosphocholine


O-1270 corresponds to PAF (C₁₆), O-1355 to PAF (C₁₈).


Synonym(s): Dehydro-PAF (C₁₈)#1-O-Oleyl-2-O-acetyl-sn-glycero-3-phosphocholine

Formula: C₂₈H₅₆NO₇P
Storage Temperature: Freezer
MDL Number: MFCD00063537
CAS Number: 85966-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR