Hexa-D-arginine

Supplier: BACHEM AMERICAS INC

Synonyms: Furin Inhibitor II

4061351.0001 4061351.0005
N-2000.0001BAEA 250.43 CAD
N-2000.0001BA N-2000.0005BA
Hexa-D-arginine
Hexa-D-arginine

Sequence: H-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-NH₂
Synonym(s): Hexa-D-arginine amide


Hexa-D-arginine amide is a potent furin inhibitor with a Ki of 0.106 μM. It also inhibited the related convertases PACE4 and prohormone convertase-1 (PC1) with Ki values of 0.58 μM and 13 μM, respectively. The stable hexapeptide exhibited protection against anthrax toxemia in vitro and in vivo.

Formula: C₃₆H₇₅N₂₅O₆
MW: 954.165 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18633383
CAS Number: 673202-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR