Z-Val-Asp(OMe)-Val-Ala-DL-Asp(OMe)-Fluoromethyl Ketone
Supplier: BACHEM AMERICAS INC
4042557.0001 4042557.0005
N-1940.0001BAEA 903.62 CAD
N-1940.0001BA N-1940.0005BA
Z-Val-Asp(OMe)-Val-Ala-DL-Asp(OMe)-Fluoromethyl Ketone
Proteins and Peptides
Methyl ester of Z-VDVAD-FMK. This cell-permeable fluoromethylketone inhibits specifically caspase-2 and, to a lesser degree, caspase-3 and caspase-7.