Fluorescein-6-carbonyl-Asp(OMe)-Glu(OMe)-Val-DL-Asp(OMe)-fluoromethylketone
Supplier: BACHEM AMERICAS INC
4045855.0500
4045855.1000
N-1905.0500BAEA
1448.37
CAD
N-1905.0500BA
N-1905.1000BA
Fluorescein-6-carbonyl-Asp(OMe)-Glu(OMe)-Val-DL-Asp(OMe)-fluoromethylketone
Proteins and Peptides
6-FAM-D(OMe)E(OMe)VD(OMe)-FMK (methyl ester of FAM-DEVD-FMK) is a cell-permeable, non-toxic inhibitor that binds irreversibly to activated caspase-3 in apoptotic cells. The fluorescence intensity can be measured by flow cytometry, microwell plate reader, or fluorescence microscopy.