L-trans-Epoxysuccinyl-Leu-4-guanidinobutylamide
Supplier: BACHEM AMERICAS INC
4027910.0005
4027910.0025
N-1645.0005BAEA
464.74
CAD
N-1645.0005BA
N-1645.0025BA
L-trans-Epoxysuccinyl-Leu-4-guanidinobutylamide
Proteins and Peptides
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).
Most commonly used diagnostic inhibitor for the characterization of cysteine proteinases. Originally isolated from cultures of Aspergillus japonicus, E64 is a non-competitive, irreversible inhibitor of thiol proteinases of the papain and calpain families.