Z-Tyr-Val-Ala-DL-Asp-Fluoromethylketone
Supplier: BACHEM AMERICAS INC
4027532.0001
4027532.0005
N-1595.0001BAEA
513.75
CAD
N-1595.0001BA
N-1595.0005BA
Z-Tyr-Val-Ala-DL-Asp-Fluoromethylketone
Proteins and Peptides
Z-YVAD-FMK, a cell-permeable potent irreversible inhibitor of caspase-1.