Z-Val-Ala-DL-Asp(OMe)-FluoromethylKetone
Supplier: BACHEM AMERICAS INC
4027403.0001
4027403.0005
N-1560.0001BAEA
317.74
CAD
N-1560.0001BA
N-1560.0005BA
Z-Val-Ala-DL-Asp(OMe)-FluoromethylKetone
Proteins and Peptides
See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).
Z-VAD(OMe)-FMK, a pan-caspase inhibitor, is a competitive irreversible peptide inhibitor and blocks caspase-1 family and caspase-3 family enzymes. The methyl ester of Z-VAD-FMK has been used in studies to support the hypothesis that inhibitors of caspases can limit brain infarction resulting from the permanent obstruction of a brain artery.