Z-Asp(OMe)-Glu(OMe)-Val-DL-Asp(OMe)-Fluoromethylketone
Supplier: BACHEM AMERICAS INC
4027402.0001
4027402.0005
N-1555.0001BAEA
407.1
CAD
N-1555.0001BA
N-1555.0005BA
Z-Asp(OMe)-Glu(OMe)-Val-DL-Asp(OMe)-Fluoromethylketone
Proteins and Peptides
This specific caspase-3 inhibitor reduces vulnerability to the neuronal death that occurs in the aftermath of kainic acid-evoked status epilepticus (SE).