AC1NUZLU (synthetic)

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Val-Ala-DL-Asp-fluoromethylketone

4026865.0005 4026865.0025
N-1510.0005BAEA 904.68 CAD
N-1510.0005BA N-1510.0025BA
AC1NUZLU (synthetic)
AC1NUZLU

Sequence: Z-Val-Ala-DL-Asp-fluoromethylketone
Synonym(s): Z-VAD-FMK


See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).


Inhibited apoptosis induced by diverse stimuli. Preincubation of cells with this caspase-1 (ICE) inhibitor (50 µM) almost completely inhibited the formation of large kilobase-pair fragments of DNA, a characteristic biochemical change associated with apoptosis. Z-VAD-FMK was shown to irreversibly inhibit most types of caspases, hence it is also called pan-caspase inhibitor. For a cell-permeable analog see N-1650, Inhibitor of peptide:N-glycanase (PNGase).

Formula: C₂₁H₂₈FN₃O₇
MW: 453.467 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06795862
CAS Number: 220644-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR