Dec-RVKR-CMK

Supplier: BACHEM AMERICAS INC

4026850.0005 4026850.0025
N-1505.0005BAEA 1807.14 CAD
N-1505.0005BA N-1505.0025BA
Dec-RVKR-CMK
Dec-RVKR-CMK

Sequence: Decanoyl-Arg-Val-Lys-Arg-chloromethylketone
Synonym(s): Furin Inhibitor I#Dec-RVKR-CMK#Furin Convertase Inhibitor (Chloromethylketone)


Dec-RVKR-CMK binds to the catalytic site of furin and therefore acts as a potent inhibitor of cleavage and fusion activity of viral glycoproteins, as well as virus replication. Furthermore it was demonstrated that this compound decreased the production of β-amyloid-induced active matrix metalloproteinase 2 in U87 cells. Apart from furin, N-1505 also inhibited the proprotein convertases PC7/LPC, PACE4, and PC6 (W.Garten, personal communication).

Formula: C₃₄H₆₆ClN₁₁O₅
Storage Temperature: Freezer
MDL Number: MFCD18910843
CAS Number: 150113-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR