Z-Asp-2,6-dichlorobenzoyloxymethylketone

Supplier: BACHEM AMERICAS INC

4026406.0100 4026406.0250
N-1445.0100BAEA 925.93 CAD
N-1445.0100BA N-1445.0250BA
Z-Asp-2,6-dichlorobenzoyloxymethylketone
Z-Asp-2,6-dichlorobenzoyloxymethylketone

Sequence: Z-Asp-2,6-dichlorobenzoyloxymethylketone
Synonym(s): Z-Asp-CH₂-DCB


See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).


Z-Asp-CH2-DCB, inhibitor of caspase-1 (ICE) has been found to prevent apoptosis caused by various types of cytotoxic agents. Such a modulation of apoptotic cell death caused by antitumor agents might offer new antitumor strategies.

Formula: C₂₀H₁₇Cl₂NO₇
Storage Temperature: Freezer
MDL Number: MFCD00674885
CAS Number: 153088-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR