Amastatin · HCl
Supplier: BACHEM AMERICAS INC
4025800.0001
4025800.0005
N-1410.0001BAEA
206.44
CAD
N-1410.0001BA
N-1410.0005BA
Amastatin · HCl
Proteins and Peptides
Amastatin has originally been isolated from the culture filtrate of Streptomyces sp. ME98-M3 and is a competitive inhibitor of human serum aminopeptidase A (Ki = 1.1 · 10⁻⁶ M) and of pig kidney leucine aminopeptidase (Ki = 1.6 · 10⁻⁶ M).