MMP inhibitor I

Supplier: BACHEM AMERICAS INC

4025806.0005 4025806.0025
N-1405.0005BAEA 232.35 CAD
N-1405.0005BA N-1405.0025BA
MMP inhibitor I
MMP inhibitor I

Sequence: 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOH
Synonym(s): MMP Inhibitor 1


This water-soluble peptidylhydroxamic acid is an inhibitor of human matrix metalloproteinases. 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOH retains its activity even after prolonged incubation with pronase or human granulocyte elastase. It inhibits interstitial and granulocyte collagenases, granulocyte gelatinase and skin fibroblast stromelysin with IC₅₀ of 1 · 10⁻⁶ M, 3 · 10⁻⁵ M and 1.5 · 10⁻⁴ M, respectively.

Formula: C₂₃H₃₄N₆O₆
Storage Temperature: Freezer
MDL Number: MFCD18910842
CAS Number: 124168-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR